Anti-LGALS1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6594R-CY5
BOSSBS-6594R-CY5EA 922 EUR
BOSSBS-6594R-CY5
Anti-LGALS1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
May regulate apoptosis, cell proliferation and cell differentiation. Binds beta-galactoside and a wide array of complex carbohydrates. Inhibits CD45 protein phosphatase activity and therefore the dephosphorylation of Lyn kinase. Strong inducer of T-cell apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LGALS1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR