Anti-GYPC Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6622R-CY5.5
BOSSBS-6622R-CY5.5EA 900 EUR
BOSSBS-6622R-CY5.5
Anti-GYPC Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This protein is a minor sialoglycoprotein in human erythrocyte membranes. The blood group Gerbich antigens and receptors for Plasmodium falciparum merozoites are most likely located within the extracellular domain. Glycophorin-C plays an important role in regulating the stability of red cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glycophorin C
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR