Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-6624R-A350
BOSSBS-6624R-A350EA 893 EUR
BOSSBS-6624R-A350
Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR