Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-6624R-A555
BOSSBS-6624R-A555EA 893 EUR
BOSSBS-6624R-A555
Anti-IHH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat