Anti-IHH Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-6624R-CY3
BOSSBS-6624R-CY3EA 900 EUR
BOSSBS-6624R-CY3
Anti-IHH Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat