Anti-IHH Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6624R-CY5
BOSSBS-6624R-CY5EA 900 EUR
BOSSBS-6624R-CY5
Anti-IHH Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Ihh is an intercellular signalling peptide essential for a variety of patterning events during development. It binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. Ihh induces the expression of parathyroid hormone-related protein (PTHRP) and is implicated in endochondral ossification, possibly regulating the balance between growth and ossification of the developing bones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IHH
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR