Anti-EVC2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6638R
BOSSBS-6638REA 725 EUR
BOSSBS-6638R
Anti-EVC2 Rabbit Polyclonal Antibody
Antikörper
EVC2 is an integral membrane protein that plays a vital role in bone formation and skeletal development. Defects in EVC2 are a cause of Ellis-van Creveld syndrome (EVC), also known as chondroectodermal dysplasia. EVC is an autosomal recessive disorder characterized by the clinical tetrad of chondrodystrophy, polydactyly, ectodermal dysplasia and cardiac anomalies. Patients manifest short-limb dwarfism, short ribs, postaxial polydactyly and dysplastic nails and teeth. Congenital heart defects, most commonly an atrioventricular septal defect, are observed in 60% of affected individuals.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: EVC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR