Anti-Vinculin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6640R-A750
BOSSBS-6640R-A750EA 378 EUR
BOSSBS-6640R-A750
Anti-Vinculin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Actin filament (F-actin)-binding protein involved in cell-matrix adhesion and cell-cell adhesion. Regulates cell-surface E-cadherin expression and potentiates mechanosensing by the E-cadherin complex. May also play important roles in cell morphology and locomotion.

Type: Primary
Antigen: Vinculin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1001-1134/1134
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat; Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR