Anti-SCYL1BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6643R-A680
BOSSBS-6643R-A680EA 893 EUR
BOSSBS-6643R-A680
Anti-SCYL1BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Defects in GORAB are the cause of geroderma osteodysplasticum (GO) [MIM:231070]; also known as gerodermia osteodysplastica or Walt Disney dwarfism. GO is a rare autosomal recessive disorder characterised by lax, wrinkled skin, joint laxity and a typical face with a prematurely aged appearance. Skeletal signs include severe osteoporosis leading to frequent fractures, malar and mandibular hypoplasia and a variable degree of growth retardation.

Type: Primary
Antigen: SCYL1BP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/394
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR