Anti-GORAB Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6643R-CY3
BOSSBS-6643R-CY3EA 900 EUR
BOSSBS-6643R-CY3
Anti-GORAB Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Defects in GORAB are the cause of geroderma osteodysplasticum (GO) [MIM:231070]; also known as gerodermia osteodysplastica or Walt Disney dwarfism. GO is a rare autosomal recessive disorder characterized by lax, wrinkled skin, joint laxity and a typical face with a prematurely aged appearance. Skeletal signs include severe osteoporosis leading to frequent fractures, malar and mandibular hypoplasia and a variable degree of growth retardation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCYL1BP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR