Anti-GORAB Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-6643R-CY5.5
BOSSBS-6643R-CY5.5EA 900 EUR
BOSSBS-6643R-CY5.5
Anti-GORAB Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Defects in GORAB are the cause of geroderma osteodysplasticum (GO) [MIM:231070]; also known as gerodermia osteodysplastica or Walt Disney dwarfism. GO is a rare autosomal recessive disorder characterized by lax, wrinkled skin, joint laxity and a typical face with a prematurely aged appearance. Skeletal signs include severe osteoporosis leading to frequent fractures, malar and mandibular hypoplasia and a variable degree of growth retardation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SCYL1BP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat