Anti-ADORA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-6649R-HRP
BOSSBS-6649R-HRPEA 897 EUR
BOSSBS-6649R-HRP
Anti-ADORA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Adenosine is involved in a variety of processes, including the synthesis of urea, the anti-inflammatory response, and the inhibition of protein synthesis. The Adenosine receptors, including Adenosine A1-R, Adenosine A2A-R, Adenosine A2B-R and Adenosine A3-R, are integral membrane proteins that are members of the G protein-coupled receptor family. Adenosine A1-R mediates ureagenesis in a partially calcium-dependent manner. Adenosine is known to mediate coronary vasodilation via Adenosine A2A-R. Collagen synthesis and total protein synthesis are inhibited in certain cells by Adenosine, acting via the A2B receptors. Activation of Adenosine A3-R inhibits the induction of TNF?and blocks the endotoxin CD14 receptor signal transduction pathway.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR