Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-6650R-A488
BOSSBS-6650R-A488EA 893 EUR
BOSSBS-6650R-A488
Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Iron-sulfur protein (IP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat