Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6650R-A680
BOSSBS-6650R-A680EA 893 EUR
BOSSBS-6650R-A680
Anti-SDHB Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Iron-sulfur protein (IP) subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Type: Primary
Antigen: SDHB
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-280/280
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR