Anti-ADA Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-6654R-A488
BOSSBS-6654R-A488EA 893 EUR
BOSSBS-6654R-A488
Anti-ADA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. Acts as a positive regulator of T-cell coactivation, by binding DPP4. Its interaction with DPP4 regulates lymphocyte-epithelial cell adhesion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR