Anti-Adenosine deaminase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6654R-A750
BOSSBS-6654R-A750EA 893 EUR
BOSSBS-6654R-A750
Anti-Adenosine deaminase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. Acts as a positive regulator of T-cell coactivation, by binding DPP4. Its interaction with DPP4 regulates lymphocyte-epithelial cell adhesion.

Type: Primary
Antigen: Adenosine deaminase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-170/363
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR