Anti-ADA Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-6654R-CY5
BOSSBS-6654R-CY5EA 900 EUR
BOSSBS-6654R-CY5
Anti-ADA Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. Acts as a positive regulator of T-cell coactivation, by binding DPP4. Its interaction with DPP4 regulates lymphocyte-epithelial cell adhesion.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat