Anti-KCNK3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-6674R-A647
BOSSBS-6674R-A647EA 893 EUR
BOSSBS-6674R-A647
Anti-KCNK3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
KCNK3 encodes one of the members of the superfamily of potassium channel proteins containing two pore forming P domains. The gene product is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: KCNK3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat