Anti-KCNK3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6674R-CY5
BOSSBS-6674R-CY5EA 900 EUR
BOSSBS-6674R-CY5
Anti-KCNK3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
KCNK3 encodes one of the members of the superfamily of potassium channel proteins containing two pore forming P domains. The gene product is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KCNK3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR