Anti-KCNN4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-6675R-A488
BOSSBS-6675R-A488EA 915 EUR
BOSSBS-6675R-A488
Anti-KCNN4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Forms a voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization which promotes calcium influx. Required for maximal calcium influx and proliferation during the reactivation of naive T-cells. The channel is blocked by clotrimazole and charybdotoxin but is insensitive to apamin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KCNN4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat