Anti-KCNN4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-6675R-A680
BOSSBS-6675R-A680EA 915 EUR
BOSSBS-6675R-A680
Anti-KCNN4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Forms a voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarisation which promotes calcium influx. Required for maximal calcium influx and proliferation during the reactivation of naive T-cells. The channel is blocked by clotrimazole and charybdotoxin but is insensitive to apamin.
Type: Primary
Antigen: KCNN4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 325-427/427
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat