Anti-KCNN4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6675R-CY3
BOSSBS-6675R-CY3EA 915 EUR
BOSSBS-6675R-CY3
Anti-KCNN4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Forms a voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization which promotes calcium influx. Required for maximal calcium influx and proliferation during the reactivation of naive T-cells. The channel is blocked by clotrimazole and charybdotoxin but is insensitive to apamin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KCNN4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR