Anti-Uricase Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6716R
BOSSBS-6716REA 725 EUR
BOSSBS-6716R
Anti-Uricase Rabbit Polyclonal Antibody
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR