Anti-UOXP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-6716R-A555
BOSSBS-6716R-A555EA 893 EUR
BOSSBS-6716R-A555
Anti-UOXP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR