Anti-Uricase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-6716R-A680
BOSSBS-6716R-A680EA 893 EUR
BOSSBS-6716R-A680
Anti-Uricase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.
Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/303
Host: Rabbit
Isotype: IgG
Reactivity: Mouse