Anti-Uricase Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6716R-A750
BOSSBS-6716R-A750EA 893 EUR
BOSSBS-6716R-A750
Anti-Uricase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.

Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/303
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR