Anti-UOXP Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-6716R-CY5
BOSSBS-6716R-CY5EA 900 EUR
BOSSBS-6716R-CY5
Anti-UOXP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat