Anti-UOXP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-6716R-FITC
BOSSBS-6716R-FITCEA 900 EUR
BOSSBS-6716R-FITC
Anti-UOXP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Uricase catalyzes the oxidation of uric acid to 5 hydroxyisourate which spontaneously decomposes to form allantoin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR