Anti-KIRREL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-6721R-A680
BOSSBS-6721R-A680EA 893 EUR
BOSSBS-6721R-A680
Anti-KIRREL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
KIRREL2 is an immunoglobulin super family gene, it has been found to be expressed in beta cells of pancreatic islets; KIRR2_HUMAN.
Type: Primary
Antigen: KIRREL2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-260/708
Host: Rabbit
Isotype: IgG
Reactivity: Rat