Anti-KIRREL2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6721R-A750
BOSSBS-6721R-A750EA 893 EUR
BOSSBS-6721R-A750
Anti-KIRREL2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
KIRREL2 is an immunoglobulin super family gene, it has been found to be expressed in beta cells of pancreatic islets; KIRR2_HUMAN.

Type: Primary
Antigen: KIRREL2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 165-260/708
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR