Anti-KIRREL2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-6721R-CY7
BOSSBS-6721R-CY7EA 900 EUR
BOSSBS-6721R-CY7
Anti-KIRREL2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
KIRREL2 is an immunoglobulin super family gene, it has been found to be expressed in beta cells of pancreatic islets; KIRR2_HUMAN.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIRREL2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR