Anti-CAMK4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6728R-CY3
BOSSBS-6728R-CY3EA 930 EUR
BOSSBS-6728R-CY3
Anti-CAMK4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAMK4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol CAMK4
Environmentally Preferable
Antigen-Name Calcium/calmodulin-dependent Protein Kinase 4
Antigen-Synonyme CAMK|CaMK IV|CAMK-GR|CAMK4|Calcium/calmodulin-dependent protein kinase type IV|CAMKIV|CaM kinase-GR
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy3®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Isotyp IgG
ImmunoChemie Yes
Modification phospho-Thr196/Thr200
Immunogen 192-203/469
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR