Anti-CAMK4 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-6728R-CY7
BOSSBS-6728R-CY7EA 930 EUR
BOSSBS-6728R-CY7
Anti-CAMK4 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAMK4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR