Anti-SLC26A4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-6787R-A680
BOSSBS-6787R-A680EA 890 EUR
BOSSBS-6787R-A680
Anti-SLC26A4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Sodium-independent transporter of chloride and iodide.

Type: Primary
Antigen: SLC26A4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/780
Host: Rabbit
Isotype: IgG
Reactivity: Human; Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR