Anti-14-3-3 zeta/delta Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-6790R-A750
BOSSBS-6790R-A750EA 893 EUR
BOSSBS-6790R-A750
Anti-14-3-3 zeta/delta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Adapter protein implicated in the regulation of a large spectrum of both general and specialised signaling pathways. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.

Type: Primary
Antigen: 14-3-3 zeta / delta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/245
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR