Anti-MAGEB3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6823R-CY5
BOSSBS-6823R-CY5EA 900 EUR
BOSSBS-6823R-CY5
Anti-MAGEB3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
MAGEB3 is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is localized in the DSS (dosage-sensitive sex reversal) critical region. It is expressed in testis and placenta, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAGEB3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR