Anti-IER3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-6840R-CY5
BOSSBS-6840R-CY5EA 900 EUR
BOSSBS-6840R-CY5
Anti-IER3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
May play a role in the ERK signaling pathway by inhibiting the dephosphorylation of ERK by phosphatase PP2A-PPP2R5C holoenzyme. Acts also as an ERK downstream effector mediating survival. As a member of the NUPR1/RELB/IER3 survival pathway, may provide pancreatic ductal adenocarcinoma with remarkable resistance to cell stress, such as starvation or gemcitabine treatment.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IER3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR