Anti-IL4I1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-6841R-A350
BOSSBS-6841R-A350EA 893 EUR
BOSSBS-6841R-A350
Anti-IL4I1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
This gene encodes a protein with limited similarity to L-amino acid oxidase which contains the conserved amino acids thought to be involved in catalysis and binding of flavin adenine dinucleotide (FAD) cofactor. The expression of this gene can be induced by interleukin 4 in B cells, however, expression of transcripts containing the first two exons of the upstream gene is found in other cell types. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2012].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL4I1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat