Anti-IL4I1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-6841R-CY5.5
BOSSBS-6841R-CY5.5EA 900 EUR
BOSSBS-6841R-CY5.5
Anti-IL4I1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This gene encodes a protein with limited similarity to L-amino acid oxidase which contains the conserved amino acids thought to be involved in catalysis and binding of flavin adenine dinucleotide (FAD) cofactor. The expression of this gene can be induced by interleukin 4 in B cells, however, expression of transcripts containing the first two exons of the upstream gene is found in other cell types. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL4I1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR