Anti-HAUS1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-6908R-CY3
BOSSBS-6908R-CY3EA 900 EUR
BOSSBS-6908R-CY3
Anti-HAUS1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
HAUS1 is 1 of 8 subunits of the 390-kD human augmin complex, or HAUS complex. The augmin complex was first identified in Drosophila, and its name comes from the Latin verb 'augmentare,' meaning 'to increase.' The augmin complex is a microtubule-binding complex involved in microtubule generation within the mitotic spindle and is vital to mitotic spindle assembly (Goshima et al., 2008 [PubMed 18443220]; Uehara et al., 2009 [PubMed 19369198]).[supplied by OMIM, Jun 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCDC5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat