Anti-CFB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-6949R-A647
BOSSBS-6949R-A647EA 893 EUR
BOSSBS-6949R-A647
Anti-CFB Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The complement component proteins, C3, C4 and C5, are potent anaphylatoxins that are released during complement activation. Binding of these proteins to their respective G protein-coupled receptors, C3aR, C1R and C5aR, induces proinflammatory events, such as cellular degranulation, smooth muscle contraction, arachidonic acid metabolism, cytokine release, leukocyte activation and cellular chemotaxis. Complement Factor B, also designated Properdin Factor B or PBF2, is part of the alternate pathway of the complement system and is cleaved by Factor D into two fragments: Ba and Bb. Bb combinesMore Product Information