Anti-H6PD Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-6989R
BOSSBS-6989REA 725 EUR
BOSSBS-6989R
Anti-H6PD Rabbit Polyclonal Antibody
Antikörper
H6PD (hexose-6-phosphate dehydrogenase, GDH/6PGL endoplasmic bifunctional protein) is a 789 amino acid protein encoded by the human gene H6PD. The N-terminal section of H6PD belongs to the glucose-6-phosphate dehydrogenase family, while the C-terminal section belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. H6PD is responsible primarily for the oxidation of glucose-6-phosphate and glucose. It also oxidizes other hexose-6-phosphates. H6PD catalyzes the conversion of glucose 6-phosphate to 6-phosphogluconolactone within the lumen of the endoplasmic reticulum, thereby generating reduced nicotinamide adenine dinucleotide phosphate. Reduced nicotinamide adenine dinucleotide phosphate is a necessary cofactor for the reductase activity of 11∫-hydroxysteroid dehydrogenase type 1, which converts hormonally inactive cortisone to active cortisol (in rodents, 11-dehydrocorticosterone to corticosterone).

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: G6PDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR