Anti-LAMTOR4 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-6991R-CY3
BOSSBS-6991R-CY3EA 900 EUR
BOSSBS-6991R-CY3
Anti-LAMTOR4 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
In mammalian cells, transcription is regulated in part by high molecular weight coactivating complexes that mediate signaling between transcriptional activators and initiation factors. These complexes include the thyroid hormone receptor-associated protein (TRAP) complex, which interacts with thyroid receptors (TR), vitamin D receptors and other steroid receptors to facilitate hormone induced transcriptional activation. The TRAP complex consists of numerous proteins ranging in size including TRAP95, TRAP100, TRAP150, TRAP220 and TRAP230, that are characterized by the presence of a nuclear receptor recognition motif which mediates the ligand-dependent binding of TRAP proteins to the nuclear receptors. TRAP220 and TRAP100 are widely expressed and most abundantly detected in skeletal muscle, heart and placenta. TRAP95, TRAP150 and TRAP230 facilitate TR induced transcription by associating with an additional transcriptional coactivating complex SMCC (SRB and MED protein cofactor complex), which consists of various subunits that share homology with several components of the yeast transcriptional mediator complexes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAMTOR4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR