Anti-ELOVL5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-7054R-FITC
BOSSBS-7054R-FITCEA 900 EUR
BOSSBS-7054R-FITC
Anti-ELOVL5 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Condensing enzyme that catalyzes the synthesis of monounsaturated and of polyunsaturated very long chain fatty acids Acts specifically toward polyunsaturated acyl-CoA with the higher activity toward C18:3(n-6) acyl-CoA.Tissue specificity: Ubiquitous. Highly expressed in the adrenal gland and testis. Weekly expressed in prostate, lung and brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELOVL5
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR