Anti-FTO Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7056R-A680
BOSSBS-7056R-A680EA 893 EUR
BOSSBS-7056R-A680
Anti-FTO Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Dioxygenase that repairs alkylated DNA and RNA by oxidative demethylation. Has highest activity towards single-stranded RNA containing 3-methyluracil, followed by single-stranded DNA containing 3-methylthymine. Has low demethylase activity towards single-stranded DNA containing 1-methyladenine or 3-methylcytosine. Specifically demethylates N(6)-methyladenosine (m6A) RNA, the most prevalent internal modification of messenger RNA (mRNA) in higher eukaryotes. Has no activity towards 1-methylguanine. Has no detectable activity towards double-stranded DNA. Requires molecular oxygen, alpha-ketoglutarate and iron. Contributes to the regulation of the global metabolic rate, energy expenditure and energy homeostasis. Contributes to the regulation of body sise and body fat accumulation.
Type: Primary
Antigen: FTO
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 165-265/505
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat