Anti-NT5C1A Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7060R-CY5
BOSSBS-7060R-CY5EA 900 EUR
BOSSBS-7060R-CY5
Anti-NT5C1A Rabbit Polyclonal Antibody (Cy5®)
Antikörper
NT5C1A dephosphorylates the 5' and 2'(3')-phosphates of deoxyribonucleotides and has a broad substrate specificity. It helps to regulate adenosine levels in the heart during ischemia and hypoxia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NT5C1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR