Anti-GADD45G Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-7069R-A488
BOSSBS-7069R-A488EA 890 EUR
BOSSBS-7069R-A488
Anti-GADD45G Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45G is highly expressed in placenta.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GADD45 gamma
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR