Anti-CARD9 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-7089R-CY3
BOSSBS-7089R-CY3EA 900 EUR
BOSSBS-7089R-CY3
Anti-CARD9 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Activates NF-kappa-B via BCL10.Tissue specificity; Highly expressed in spleen. Also detected in liver, placenta, lung, peripheral blood leukocytes and in brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CARD9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR