Anti-CCDC139 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7102R
BOSSBS-7102REA 725 EUR
BOSSBS-7102R
Anti-CCDC139 Rabbit Polyclonal Antibody
Antikörper
Pseudouridination is the isomerization of uridine to pseudouridine. It is the most common posttranscriptional nucleotide modification found in RNA. It is essential for biologic functions such as spliceosome biogenesis. Pseudouridylate synthases, such as PUS10, catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. These enzymes also act as RNA chaperones which facilitate the correct folding and assembly of tRNAs.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CCDC139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR