Anti-CCDC139 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-7102R-A488
BOSSBS-7102R-A488EA 893 EUR
BOSSBS-7102R-A488
Anti-CCDC139 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Pseudouridination is the isomerization of uridine to pseudouridine. It is the most common posttranscriptional nucleotide modification found in RNA. It is essential for biologic functions such as spliceosome biogenesis. Pseudouridylate synthases, such as PUS10, catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. These enzymes also act as RNA chaperones which facilitate the correct folding and assembly of tRNAs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC139
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR